Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: YTDSSYEEKK YTMGDAPDYD RSQWLNEKFK LGLDFPNLPY LIDGAHKITQ SNAILCYIAR KHNLCGETEE EKIRVDILEN QAMDVSNQLA RVCYSPD
Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-transferase that belongs to the mu class. The mu class of enzymes functions in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding the mu class of enzymes are organized in a gene cluster on chromosome 1p13.3 and are known to be highly polymorphic. These genetic variations can change an individual's susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of certain drugs. Diversification of these genes has occurred in regions encoding substrate-binding domains, as well as in tissue expression patterns, to accommodate an increasing number of foreign compounds. Multiple transcript variants, each encoding a distinct protein isoform, have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glutathione S-alkyltransferase M4; glutathione S-aralkyltransferase M4; glutathione S-aryltransferase M4; glutathione S-transferase M4; Glutathione S-transferase Mu 4; GST class-mu 4; GST-Mu2; GSTM4-4; GTS-Mu2; Leukotriene C4 synthase GSTM4; S-(hydroxyalkyl)glutathione lyase M4; testis tissue sperm-binding protein Li 60n
Gene Aliases: GSTM4; GSTM4-4; GTM4
UniProt ID: (Human) Q03013
Entrez Gene ID: (Human) 2948
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support