Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MSGDATRTLG KGSQPPGPVP EGLIRIYSMR FCPYSHRTRL VLKAKDIRHE VVNINLRNKP EWYYTKHPFG HIPVLETSQC QLIYESVIAC EYLDDAYPGR KLFPYDPYER ARQKMLLELF CKVPHLTKEC LVALRCGREC TNLKAALRQE FSNLEEILEY QNTTFFGGTC ISMIDYLLWP WFERLDVYGI LDCVSHTPAL RLWISAMKWD PTVCALLMDK SIFQGFLNLY FQNNPNAFDF GLC
The omega class glutathione transferases (GST; EC 2.5.1.18) have poor activity with common GST substrates, but exhibit novel glutathione-dependent thioltransferase, dehydroascorbate reductase, and monomethylarsonate reductase activities, and they modulate Ca(2+) release by ryanodine receptors (e.g., RYR1).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA127L20.1 (novel glutathione-S-transferase); Glutathione S-transferase omega 2-2; Glutathione S-transferase omega-2; Glutathione-dependent dehydroascorbate reductase; glutathione-S-transferase-like protein; GSTO 2-2; GSTO-2; MMA(V) reductase; Monomethylarsonic acid reductase
Gene Aliases: bA127L20.1; GSTO 2-2; GSTO2
UniProt ID: (Human) Q9H4Y5
Entrez Gene ID: (Human) 119391
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support