Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Reacts specifically with S1 protein. Cross reactivity from other sources has not been determined.
GST (Glutathione S-transferase) is a 26kDa protein, present in eukaryotes and in prokaryotes, where they catalyze a variety of reactions. Glutathione Stransferase is used to create the GST gene fusion system. This GST-fusion protein can then be purified from cells via its high affinity for glutathione. Fusion Proteins offer an important biological assay for direct protein-to-protein interactions. The GST gene fusion system has been introduced into numerous expression vectors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activator protein 1; AP1; Cancer/testis antigen 48; CT48; deafness, X-linked 7; enhancer-binding protein AP1; epididymis secretory protein Li 22; fatty acid ethyl ester synthase III; glutathione S-alkyltransferase; glutathione S-aralkyltransferase; glutathione S-aryltransferase; glutathione S-transferase M1; Glutathione S-transferase Mu 1; Glutathione S-transferase P; Glutathione-S-Transferase; Gonad-specific transporter; GST; GST class-mu 1; GST class-pi; GST HB subunit 4; GST protein; GST Tag; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GSTP1-1; GTH4; HB subunit 4; Jun activation domain binding protein; jun oncogene; jun proto-oncogene; OATP-I; Organic anion-transporting polypeptide 6A1; Organic anion-transporting polypeptide I; p39; Proto-oncogene c-Jun; S-(hydroxyalkyl)glutathione lyase; Solute carrier family 21 member 19; Solute carrier organic anion transporter family member 6A1; solute carrier organic anion transporter family, member 6A1; testicular tissue protein Li 174; testis-specific organic anion transporter; Transcription factor AP-1 subunit Jun; Transcription factor Jun; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog
Gene Aliases: AP-1; AP1; c-Jun; CT48; DFN7; FAEES3; GST; GST1; GST3; GSTM1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GSTP; GSTP1; GTH4; GTM1; H-B; HEL-S-22; JUN; MU; MU-1; OATP-I; OATP6A1; OATPY; PI; SLC21A19; SLCO6A1
UniProt ID: (Human) P09488, (Human) P09211, (Human) P05412, (Human) Q86UG4
Entrez Gene ID: (Human) 2944, (Human) 2950, (Human) 3725, (Human) 133482
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support