Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGLELYLDLL SQPCRAVYIF AKKNDIPFEL RIVDLIKGQH LSDACAQVNP LKKVPALKDG DFTLTESVAI LLYLTRKYKV PDYWYPQDLQ ARARVDEYLA WQHTTLRRSC LRALWHKVMF PVFLGEPVSP QTLAATLAEL DVTLQLLEDK FLQNKAFLTG PHISLADLVA ITELMHPVGA GCQVFEGRPK LATWRQRVEA AVGEDLFQEA HEVILKAKDF PPADPTIKQK LMPWVLAMIR
Glutathione S-transferase (GST) theta 1 (GSTT1) is a member of a superfamily of proteins that catalyze the conjugation of reduced glutathione to a variety of electrophilic and hydrophobic compounds. Human GSTs can be divided into five main classes: alpha, mu, pi, theta, and zeta. The theta class includes GSTT1 and GSTT2. The GSTT1 and GSTT2 share 55% amino acid sequence identity and both of them were claimed to have an important role in human carcinogenesis. The GSTT1 gene is located approximately 50kb away from the GSTT2 gene. The GSTT1 and GSTT2 genes have a similar structure, being composed of five exons with identical exon/intron boundaries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glutathione S-transferase theta-1; Glutathione transferase T1-1; GST class-theta-1
Gene Aliases: GSTT1
UniProt ID: (Human) P30711
Entrez Gene ID: (Human) 2952
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support