Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This gene is a member of the glutathione S-transferase (GSTs) super-family which encodes multifunctional enzymes important in the detoxification of electrophilic molecules, including carcinogens, mutagens, and several therapeutic drugs, by conjugation with glutathione. This enzyme also plays a significant role in the catabolism of phenylalanine and tyrosine. Thus defects in this enzyme may lead to severe metabolic disorders including alkaptonuria, phenylketonuria and tyrosinaemia. Several transcript variants of this gene encode multiple protein isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glutathione S-alkyltransferase; glutathione S-aralkyltransferase; glutathione S-aryltransferase; Glutathione S-transferase zeta 1; glutathione transferase zeta 1; GSTZ1-1; MAAI; Maleylacetoacetate isomerase; maleylacetone isomerase; S-(hydroxyalkyl)glutathione lyase
Gene Aliases: GSTZ1; GSTZ1-1; MAAI; MAI
UniProt ID: (Human) O43708
Entrez Gene ID: (Human) 2954
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support