Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GRTLPSFTTA ELGTSNSSAN FTFPGYPIHV PAGVTLQTVS GHLYKVNVPI MVTETS
Antigen sequence identity: Mouse 73%; Rat 71%
The assembly and stability of the RNA polymerase II transcription pre-initiation complex on a eukaryotic core promoter involve the effects of TFIIA on the interaction between TATA-binding protein (TBP) and DNA. This gene encodes a germ cell-specific counterpart of the large (alpha/beta) subunit of general transcription factor TFIIA that is able to stabilize the binding of TBP to DNA and may be uniquely important to testis biology. Alternative splicing for this locus has been observed and two variants, encoding distinct isoforms, have been identified. Co-transcription of this gene and the neighboring upstream gene generates a rare transcript (SALF), which encodes a fusion protein comprised of sequence sharing identity with each individual gene product.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CXCDC1; General transcription factor II A, 1-like factor; general transcription factor IIA 1 like; general transcription factor IIA 1-like; general transcription factor IIA, 1-like; GTF2A1-like factor; KIAA2037; testis secretory sperm-binding protein Li 230m; TFIIA large subunit isoform ALF; TFIIA-alpha and beta-like factor; TFIIA-alpha/beta-like factor
Gene Aliases: ALF; GTF2A1L; GTF2A1LF
UniProt ID: (Human) Q9UNN4
Entrez Gene ID: (Human) 11036
Molecular Function: general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support