Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TFIIF is a general transcription initiation factor that binds to RNA polymerase II and helps to recruit it to the initiation complex in collaboration with TFIIB. It promotes transcription elongation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: General transcription factor IIF 74 kDa subunit; general transcription factor IIF polypeptide 1 (74kD subunit); General transcription factor IIF subunit 1; general transcription factor IIF, polypeptide 1, 74kDa; MGC94148; OTTHUMP00000237859; TFIIF-alpha; Transcription initiation factor IIF subunit alpha; Transcription initiation factor RAP74
Gene Aliases: 2810405L04Rik; BTF4; C76800; GTF2F1; RAP74; TF2F1; TFIIF
UniProt ID: (Human) P35269, (Mouse) Q3THK3
Entrez Gene ID: (Human) 2962, (Mouse) 98053
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support