Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Positive Control: Rat testis tissue lysate, A549 cell lysate, HepG2 cell lysate, HL-60 cell lysate, THP-1 cell lysate, Daudi cell lysate, rat testis tissue, human skin tissue, human placenta tissue, human uterus tissue.
Subcellular Location: Nucleus.
GTF2F2 is required for both basal and HIV-1 Tat-activated transcription. GTF2F2 synergizes with HIV-1 Tat and the cellular co-activator Tat-SF1 during Tat-mediated transactivation of the HIV-1 LTR promoter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent helicase GTF2F2; General transcription factor IIF 30 kDa subunit; General transcription factor IIF subunit 2; general transcription factor IIF, polypeptide 2 (30kD subunit); general transcription factor IIF, polypeptide 2, 30kDa; TFIIF beta; TFIIF-beta; transcription factor gamma; Transcription initiation factor IIF subunit beta; Transcription initiation factor RAP30
Gene Aliases: BTF4; GTF2F2; RAP30; TF2F2; TFIIF
UniProt ID: (Human) P13984
Entrez Gene ID: (Human) 2963, (Rat) 81674
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support