Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SKMITFIDL AGHHKYLHTT IFGLTSYCPD CALLLVSANT GIAGTTREHL GLALALKVPF FIVVSKIDLC AKTTVERTVR QLERVLKQPG CHKVPMLVTS EDDAVTAAQQ FAQSPNVTPI FTLSSVSGES LDLLKVFLNI LPPLTNSKEQ EELMQQLTEF QVDEIYTVPE VGTVVGGTLS SGICREGDQL VVGPTDDGCF LELRVCSIQR NRSACRVLRA GQAATLALGD FDRALLRKGM VMLSPEMNPT ICSVFEAEIV LLFHATTFRR GFQVTVHVGN VRQTAVVEKI HAKDKLRTGE KAVVRFRFLK HPEYLKVGAK LLFREGVTKG IGHVTDVQAI TAGEAQANMG F (183-532 aa encoded by BC028347)
Small G proteins act as molecular switches for regulation of variety of cellular processes, such as nuclear transport, signal transduction, membrane trafficking and protein synthesis. GTPBP2 (GTP-binding protein 2) is a 602 amino acid G protein that is expressed in kidney, skeletal muscle, testis, brain and thymus, though it is not detected in liver. Expression of GTPBP2 is enhanced by ©-interferon stimulation in HeLa cells, THP-1 cells and thioglycollate-elicited mouse peritoneal macrophages. There are four isoforms of GTPBP2 that are expressed as a result of alternative splicing events. Since mutation of the gene encoding GTPBP1 does not lead to any phenotypic abnormalities, it is thought that there may be a genetic redundancy to make up for GTPBP1 lack-of-function. GTPBP2 shares 44% sequence similarity with GTPBP1 and also overlaps in expression pattern, suggesting that the GTPBP2 gene may compensate for GTPBP1 genetic abnormalities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP-binding protein 2; GTP-binding-like protein 2; GTPBP2
Gene Aliases: GTPBP2; nmf205
UniProt ID: (Human) Q9BX10, (Mouse) Q3UJK4
Entrez Gene ID: (Human) 54676, (Mouse) 56055, (Rat) 363195
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support