Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MWRGLWTLAA QAARGPRRLC TRRSSGAPAP GSGATIFALS SGQGRCGIAV IRTSGPASGH ALRILTAPRD LPLARHASLR LLSDPRSGEP LDRALVLWFP GPQSFTGEDC VEFHVHGGPA VVSGVLQALG SVPGLRPAEA GEFTRRAFAN GKLNLTEVEG LADLIHAETE AQRRQALRQL DGELGHLCRG WAETLTKALA HVEAYIDFGE DDNLEEGVLE QADIEVRALQ VALGAHLRDA RRGQRLRSGA HVVVTGPPNA GKSSLVNLLS RKPVSIVSPE PGTTRDVLET PVDLAGFPVL LSDTAGLREG VGPVEQEGVR RARERLEQAD LILAMLDASD LASPSSCNFL ATVVASVGAQ SPSDSSQRLL LVLNKSDLLS PEGPGPGPDL PPHLLLSCLT GEGLDGLLEA LRKELAAVCG DPSTDPPLLT RARHQHHLQG CLDALGHYKQ SKDLALAAEA LRVARGHLTR LTGGGGTEEI LDIIFQDFCV GK
This locus encodes a GTP-binding protein. The encoded protein is localized to the mitochondria and may play a role in mitochondrial tRNA modification. Polymorphisms at this locus may be associated with severity of aminoglycoside-induced deafness, a disease associated with a mutation in the 12S rRNA. Alternatively spliced transcript variants encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP binding protein 3 (mitochondrial); GTP-binding protein 3; Mitochondrial GTP-binding protein 1; tRNA modification GTPase GTPBP3, mitochondrial
Gene Aliases: COXPD23; GTPBG3; GTPBP3; MSS1; MTGP1; THDF1
UniProt ID: (Human) Q969Y2, (Rat) Q5PQQ1
Entrez Gene ID: (Human) 84705, (Rat) 290633
Molecular Function:
RNA methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support