Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LPGKKLLSE KKLKRYFVDY RRVLVCGGNG GAGASCFHSE PRKEFGGPDG GDGGNGGHVI LRVDQQVKSL SSVLSRYQGF SGEDGGSKNC FGRSGAVLYI RVPVGTLVKE GGRVVADLSC VGDEYIAALG GAGGKGNRFF LANNNRAPVT CTPGQPGQQR VLHLELKTVA HAGMVGFPNA GKSSLLRAIS NARPAVASYP FTTLKPHVGI VHYEGHLQIA VADIPGIIRG AHQNRGLGSA FLRHIERCRF LLFVVDLSQP EPWTQVDDLK YELEMYEKGL SARPHAIVAN KIDLPEAQAN LSQLRDHLGQ EVIVLSALTG ENLEQLLLHL KVLYDAYAEA ELGQGRQPLR W (57-406 aa encoded by BC036716)
Small G proteins, such as GTPBP5, act as molecular switches that play crucial roles in the regulation of fundamental cellular processes such as protein synthesis, nuclear transport, membrane trafficking, and signal transduction (Hirano et al., 2006 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP binding protein 5; GTP binding protein 5 (putative); GTP-binding protein 5; GTP-binding protein 5 (putative); Mitochondrial ribosome-associated GTPase 2; ObgH1; Protein obg homolog 1
Gene Aliases: 1810011P19Rik; 2900056P18Rik; D2Bwg0647e; dJ1005F21.2; GTPBP5; MTG2; OBGH1
UniProt ID: (Human) Q9H4K7
Entrez Gene ID: (Human) 26164, (Mouse) 52856, (Rat) 296462
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support