Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MEPEAFEICP YDPHHRIPLS RFQYHLASCR RKNPKKAKKM ATCKYNACHV VPIKNLEEHE AVCVNRSAVE EEDTENPLKV SPPSSEQNDD TQQVSPCLPS PDIWNVDGAN CQHVFVLKTF FPQKVVCEND TKESARETSP QKILRPGQ
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 112, member A; gametocyte specific factor 1-like; Gametocyte-specific factor 1-like; Protein FAM112A
Gene Aliases: C20orf65; dJ1028D15.4; FAM112A; GTSF1L
UniProt ID: (Human) Q9H1H1
Entrez Gene ID: (Human) 149699
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support