Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGNGKSIAGD QKAVPTQETH VWYRTFMMEY PSGLQTLHEF KTLLGLQGLN QKANKHIDQV YNTFDTNKDG FIDFLEFIAA VNLIMQEKME QKLKWYFKLY DADGNGSIDK NELLDMFMAV QALNGQQTLS PEEFINLVFH KIDINNDGEL TLEEFINGMA KDQDLLEIVY KSFDFSNVLR VICNGKQPDM ETDSSKSPDK AGLGKVKMK
Guanylyl cyclase-activating protein 3
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GCAP 3; Guanylate cyclase activator 1C; Guanylyl cyclase-activating protein 3; Hu-DF1-VAR
Gene Aliases: GCAP3; GUCA1C
UniProt ID: (Human) O95843
Entrez Gene ID: (Human) 9626
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support