Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGCRAASGLL PGVAVVLLLL LQSTQSVYIQ YQGFRVQLES MKKLSDLEAQ WAPSPRLQAQ SLLPAVCHHP ALPQDLQPVC ASQEASSIFK TLRTIANDDC ELCVNVACTG CL
Endogenous activator of intestinal guanylate cyclase. It stimulates this enzyme through the same receptor binding region as the heat-stable enterotoxins. May be a potent physiological regulator of intestinal fluid and electrolyte transport. May be an autocrine/paracrine regulator of intestinal salt and water transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Guanylate cyclase activator 2B; guanylate cyclase activator 2B (uroguanylin); prepro-uroguanylin; uroguanylin
Gene Aliases: GCAP-II; GUCA2B; UGN
UniProt ID: (Human) Q16661
Entrez Gene ID: (Human) 2981
Molecular Function:
protein-binding activity modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support