Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EDFTGRGLY LSDIPIHNAL RDVVLIGEQA RAQDGLKKRL GKLKATLEQA HQALEEEKKK TVDLLCSIFP CEVAQQLWQG QVVQAKKFSN VTMLFSDIVG FTAICSQCSP LQVITMLNAL YTRFDQQCGE LDVYKVETIG DAYCVAGGLH KESDTHAVQI ALMAVKMMEL SDEVMSPHGE PIKMRIGLHS GSVFAGVVGV KMPRYCLFGN NVTLANKFES CSVPRKINVS PTTYRLLKDC PGFVFTPRSR EELPPNFPSE IPGICHFLDA YQQGTNSKPC FQKKDVEDGN ANFLGKASGI D (391-690 aa encoded by BC028384)
Guanylate cyclases belong to the adenylyl cyclase class-4/guanylyl cyclase family. There are two forms of guanylate cyclase, a soluble form (GCS or sGC), which act as receptors for nitric oxide and a membrane-bound receptor form (GC), which are peptide hormone receptors. The GC-C protein is composed of an extracellular domain, a single transmembrane domain, and a cytoplasmic region consisting of a kinase-like domain and a catalytic domain. It is expressed as two differentially glycosylated forms, a 130 kDa precursor form present in the endoplasmic reticulum and a 145 kDa form present on the plasma membrane. Ligand binding to the extracellular domain of GC-C promotes the accumulation of cGMP. GC-C acts as the receptor for heatstable enterotoxins, small peptides secreted by some pathogenic strains of E. coli that cause severe secretory diarrhea. GC-C also binds to guanylin and uroguanylin peptides, which modulate renal function in response to oral salt load.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 4.6.1.2; alpha 1 sGC; GC-S-alpha-1; GCS-alpha-1; GCS-alpha-3; GCS-alpha1; GCSalpha1; guanylate cyclase 1, soluble, alpha 3; Guanylate cyclase soluble alpha 1 (GTP pyrophosphate - lyase); Guanylate cyclase soluble subunit alpha-1; Guanylate cyclase soluble subunit alpha-3; Guanylate cyclase, soluble, alpha 1 (GTP pyrophosphate - lyase); GUCY1A1; Soluble guanylate cyclase large subunit
Gene Aliases: 1200016O07Rik; GC-SA3; GUC1A3; GUCA3; GUCSA3; GUCY1A1; GUCY1A3; MYMY6; SGC; sGC-alpha1
UniProt ID: (Human) Q02108, (Mouse) Q9ERL9
Entrez Gene ID: (Human) 2982, (Mouse) 60596, (Rat) 497757
Molecular Function:
guanylate cyclase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support