Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MFCTKLKDLK ITGECPFSLL APGQVPNESS EEAAGSSESC KATMPICQDI PEKNIQESLP QRKTSRSRVY LHTLAESICK LIFPEFERLN VALQRTLAKH KIKESRKSLE REDFEKTIAE QAVAAGVPVE VIKESLGEEV FKICYEEDEN ILGVVGGTLK DFLNSFSTLL KQSSHCQEAG KRGRLEDASI LCLDKEDDFL HVYYFFPKRT TSLILPGIIK AAAHVLYETE VEVSLMPPCF HNDCSEFVNQ PYLLYSVHMK STKPSLSPSK PQSSLVIPTS LFCKTFPFHF MFDKDMTILQ FGNGIRRLMN RRDFQGKPNF EEYFEILTPK INQTFSGIMT MLNMQFVVRV RRWDNSVKKS SRVMDLKGQM IYIVESSAIL FLGSPCVDRL EDFTGRGLYL SDIPIHNALR DVVLIGEQAR AQDGLKKRLG KLKATLEQAH QALEEEKKKT VDLLCSIFPC EVAQQLWQGQ VVQAKKFSNV TMLFSDIVGF TAICSQCSPL QVITMLNALY TRFDQQCGEL DVYKVETIGD AYCVAGGLHK ESDTHAVQIA LMAVKMMELS DEVMSPHGEP IKMRIGLHSG SVFAGVVGVK MPRYCLFGNN VTLANKFESC SVPRKINVSP TTYRLLKDCP GFVFTPRSRE ELPPNFPSEI PGICHFLDAY QQGTNSKPCF QKKDVEDGNA NFLGKASGID
Guanylate cyclases belong to the adenylyl cyclase class-4/guanylyl cyclase family. There are two forms of guanylate cyclase, a soluble form (GCS or sGC), which act as receptors for nitric oxide and a membrane-bound receptor form (GC), which are peptide hormone receptors. The GC-C protein is composed of an extracellular domain, a single transmembrane domain, and a cytoplasmic region consisting of a kinase-like domain and a catalytic domain. It is expressed as two differentially glycosylated forms, a 130 kDa precursor form present in the endoplasmic reticulum and a 145 kDa form present on the plasma membrane. Ligand binding to the extracellular domain of GC-C promotes the accumulation of cGMP. GC-C acts as the receptor for heatstable enterotoxins, small peptides secreted by some pathogenic strains of E. coli that cause severe secretory diarrhea. GC-C also binds to guanylin and uroguanylin peptides, which modulate renal function in response to oral salt load.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 4.6.1.2; GC-S-alpha-1; GCS-alpha-1; GCS-alpha-3; GCS-alpha1; GCSalpha1; guanylate cyclase 1, soluble, alpha 3; Guanylate cyclase soluble subunit alpha-1; Guanylate cyclase soluble subunit alpha-3; GUCY1A1; Soluble guanylate cyclase large subunit
Gene Aliases: GC-SA3; GUC1A3; GUCA3; GUCSA3; GUCY1A1; GUCY1A3; MYMY6
UniProt ID: (Human) Q02108
Entrez Gene ID: (Human) 2982
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support