Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: RKVAQGSRSS LGARSMSDIR SGPSQHLDSP NIGVYEGDRV WLKKFPGDQH IAIRPATKTA FSKLQELRHE NVALYLGLFL ARGAEGPAAL WEGNLAVVSE HCTRGSLQDL
Probably plays a specific functional role in the rods and/or cones of photoreceptors. It may be the enzyme involved in the resynthesis of cGMP required for recovery of the dark state after phototransduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CG-E; cone rod dystrophy 6; guanylate cyclase 2D, membrane (retina-specific); Guanylate cyclase 2D, retinal; RETGC-1; retinal guanylate cyclase 1; Retinal guanylyl cyclase 1; Rod outer segment membrane guanylate cyclase; ROS-GC
Gene Aliases: CORD5; CORD6; CYGD; GUC1A4; GUC2D; GUCY2D; LCA; LCA1; RCD2; RETGC; RETGC-1; RETGC1; ROS-GC1; ROSGC
UniProt ID: (Human) Q02846
Entrez Gene ID: (Human) 3000
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support