Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: CDPLSQPSPQ PADFTETETI LQPANKVESV SSEETFEPSQ ELPAEALRDP SLQDALEVDL AVSVSQISIE EKVKELSPEE ERRKWEEGRI DYMGKDAFAR IQEKLDRFLQ
GYG2 belongs to the glycogenin family. Glycogenin is a self-glycosylating protein involved in the initiation phase of glycogen biosynthesis. Multiple alternatively spliced transcript variants encoding different isoforms have been identified, including the predominant band around 66kDa and a light band around 45kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glycogenin glucosyltransferase; Glycogenin-2; GN-2
Gene Aliases: GN-2; GN2; GYG2
UniProt ID: (Human) O15488
Entrez Gene ID: (Human) 8908
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support