Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: DSPGLTFHIK AAIQHPRYKP VPALENDLAL LQLDGKVKPS RTIRPLALPS KRQVVAAGTR CSMAGWGLTH QGGRLSRVLR ELDLQVLDTR MCNNSRFWNG SLSPSMVCL
Cleaves peptide substrates after methionine, leucine, and norleucine. Physiological substrates include EZR, alpha- tubulins and the apoptosis inhibitor BIRC5/Survivin. Promotes caspase activation and subsequent apoptosis of target cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Granzyme M; granzyme M (lymphocyte met-ase 1); HU-Met-1; lymphocyte met-ase 1; Met-1 serine protease; Met-ase; Natural killer cell granular protease
Gene Aliases: GZMM; LMET1; MET1
UniProt ID: (Human) P51124
Entrez Gene ID: (Human) 3004
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support