Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Specificity: The antibody binds to galanin with an affinity constant (Kd) of 0.07 nM reported for the original antibody.
Endocrine hormone of the central and peripheral nervous systems that binds and activates the G protein-coupled receptors GALR1, GALR2, and GALR3. This small neuropeptide may regulate diverse physiologic functions including contraction of smooth muscle of the gastrointestinal and genitourinary tract, growth hormone and insulin release and adrenal secretion. [Uniprot]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Galanin peptides; galanin prepropeptide; galanin-message-associated peptide; galanin-related peptide; galanin/GMAP prepropeptide
Gene Aliases: ETL8; GAL; GAL-GMAP; GAL1; GALN; GLNN; GMAP
UniProt ID: (Human) P22466, (Mouse) P47212
Entrez Gene ID: (Human) 51083, (Mouse) 14419
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support