Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
In direct ELISAs, no cross-reactivity with recombinant human Galectin-1, -2, -3, -4, -7, -8, or -9/Ecalectin is observed.
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Regulates immune responses through the recognition of cell-surface glycans. Essential for the anergy and suppressive function of CD25-positive regulatory T-cells (Treg).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Charcot-Leyden crystal protein; CLC; EC 3.1.1.5; Eosinophil lysophospholipase; Gal-10; galectin 10; Galectin-10; lectin, galactoside-binding, soluble, 10; Lysolecithin acylhydrolase; MGC149659
Gene Aliases: CLC; Gal-10; GAL10; LGALS10; LGALS10A; LPPL_HUMAN
UniProt ID: (Human) Q05315
Entrez Gene ID: (Human) 1178
Molecular Function:
extracellular matrix protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support