Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. The protein encoded by this gene has lysophospholipase activity. It is composed of two identical subunits which are held together by disulfide bonds. This protein has structural similarity to several members of the beta-galactoside-binding S-type lectin family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta-galactoside-binding lectin; Gal-13; Galactoside-binding soluble lectin 13; Galectin-13; lectin, galactoside binding soluble 13; lectin, galactoside-binding, soluble, 13; placental protein 13; Placental tissue protein 13; PP13
Gene Aliases: GAL13; LGALS13; PLAC8; PP13
UniProt ID: (Human) Q9UHV8
Entrez Gene ID: (Human) 29124
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support