Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Positive test controls include: C2C12, Mouse lung, HeLa, A-549, RD. The target is usually found in the following locations: Cytoplasm, Secreted, cytoskeleton.
Immunogen sequence: DQTDGLGLSY LSSHIANVER VPFDAATLHT STAMAAQHGM DDDGTGQKQI WRIEGSNKVP VDPATYGQFY GGDSYIILYN YRHGGRQGQI IYNWQGAQST Q
The protein encoded by this gene binds to the "plus" ends of actin monomers and filaments to prevent monomer exchange. The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments. Defects in this gene are a cause of familial amyloidosis Finnish type (FAF). Multiple transcript variants encoding several different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actin-depolymerizing factor; ADF; ADFBrevin; AGEL; Brevin; DKFZp313L0718; Gelsolin; RP11-477J21.1
Gene Aliases: ADF; AGEL; Gsb; GSN
UniProt ID: (Human) P06396, (Mouse) P13020
Entrez Gene ID: (Human) 2934, (Mouse) 227753
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support