Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Immunogen sequence: YLYDSDRGLS TLQAVLDSAA EKKWQVTAIN VGNINNDKKD EMYRSLFQDL ELKKERRVIL DCERDKVNDI VDQVITIGKH VKGYHYIIAN LGFTDGDLLK I
The ion channels activated by glutamate are typically divided into two classes. Those that are sensitive to N-methyl-D-aspartate (NMDA) are designated NMDA receptors (NMDAR) while those activated by alpha-amino-3-hydroxy-5 -methyl-4-isoxalone propionic acid (AMPA) are known as AMPA receptors (AMPAR). The AMPAR are comprised of four distinct Glutamate Receptor Subunits designated (GluR1-4) and they play key roles in virtually all excitatory neurotransmission in the brain. The GluR2 subunit is thought to play a key role in forms of synaptic plasticity such as LTD.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AMPA selective glutamate receptor; AMPA-selective glutamate receptor 2; GluA2; GluR-2; GluR-B; GluR-K2; glutamate receptor; Glutamate receptor 2; glutamate receptor B; glutamate receptor B flip isoform; Glutamate receptor ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA 2; OTTHUMP00000165324; OTTHUMP00000220942; OTTHUMP00000220943
Gene Aliases: GluA2; Glur-2; GluR-B; GluR-K2; GLUR2; GLURB; GRIA2; HBGR2
UniProt ID: (Human) P42262, (Rat) P19491
Entrez Gene ID: (Human) 2891, (Mouse) 14800, (Rat) 29627
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support