Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Fabgennix
Glucose-6-phosphatase (G6PC) is an integral membrane protein of the endoplasmic reticulum that catalyzes the hydrolysis of D-glucose 6-phosphate to D-glucose and orthophosphate. It is a key enzyme in glucose homeostasis, functioning in gluconeogenesis and glycogenolysis. Defects in the enzyme cause glycogen storage disease type I.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G-6-Pase 2; G6Pase 2; Glucose-6-phosphatase 2; glucose-6-phosphatase, catalytic, 2; glucose-6-phosphatase, catalytic, related sequence; islet-specific G6CP-related protein; Islet-specific glucose-6-phosphatase catalytic subunit-related protein; islet-specific glucose-6-phosphatase-related protein; similar to glucose-6-phosphatase, catalytic, related
Gene Aliases: G6pc-rs; G6PC2; IGRP
UniProt ID: (Human) Q9NQR9, (Mouse) Q9Z186
Entrez Gene ID: (Human) 57818, (Mouse) 14378, (Rat) 681817
Molecular Function:
phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support