Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in rat and mouse.
Specificity of this antibody: GLRA3.
The GLRA3 gene encodes the alpha-3 subunit of the neuronal glycine receptor, a ligand-gated chloride channel composed of ligand-binding alpha and structural beta polypeptides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glycine receptor; Glycine receptor subunit alpha-3; glycine receptor, alpha 3; glycine receptor, alpha 3 subunit; glycine receptor, alpha-3 polypeptide; ligand gated ion channel
Gene Aliases: GLRA3
UniProt ID: (Human) O75311, (Mouse) Q91XP5
Entrez Gene ID: (Human) 8001, (Rat) 114516, (Mouse) 110304
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support