Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: WRNLLDSMVL ITDKFWGTSG VESVIGSVHT WLAEAINALQ DNRDTLTAKV IQGCGNPKVN PQGPGPEEKR RRGKLAPRER PPSGTLEKLV SEAKAQLRDV QDFWISLPGT LCSEKMALST ASDDRCWNGM A; Positive Samples: NIH/3T3, Mouse brain, Rat brain; Cellular Location: Cell membrane, Endosome, Extracellular side, GPI-anchor, Lipid-anchor, extracellular space
Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glypican proteoglycan 1; Glypican-1; GPI-anchored cell surface heparan sulfate proteoglycan; HSPG M12
Gene Aliases: AI462976; glypican; GPC1; HSPG M12; HSPGM12
UniProt ID: (Human) P35052, (Mouse) Q9QZF2, (Rat) P35053
Entrez Gene ID: (Human) 2817, (Mouse) 14733, (Rat) 58920
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support