Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ALNLGIEVI NTTDHLKFSK DCGRMLTRMW YCSYCQGLMM VKPCGGYCNV VMQGCMAGVV EIDKYWREYI LSLEELVNGM YRIYDMENVL LGLFSTIHDS IQYVQKNAGK LTTTIGKLCA HSQQRQYRFA YYPEDLFIDK KVLKVAHVEH EETLSSRRRE LIQKLKSFIS FYSALPGYIC SHSPVAENDT LCWNGQELVE RYSQKAARNG MKNQFNLHEL KMKGPEPVVS QIIDKLKHIN QLLRTMSMPK GRVLDKNLDE EGFESGDCGD DEDECIGGSG DGMIKVKNQL RFLAELAYDL DVDDAPGNSQ QATPKDNEIS TFHNLGNVHS PLKLLTSMAI SVVCFFFLVH (232-580 aa encoded by BC035972)
GPC3 is a cell surface proteoglycan that bears heparan sulfate. This protein may be involved in the suppression/modulation of growth in the predominantly mesodermal tissues and organs, and may play a role in the modulation of IGF2 interactions with its receptor and thereby modulate its function. Members of the glypican-related integral membrane proteoglycan family contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol (GPI) linkage. These proteins may play a role in the control of cell division, growth regulation, and tumor predisposition. Deletion mutations in GPC3 are the cause of Simpson-Golabi-Behmel syndrome (SGBS), also known as Simpson dysmorphia syndrome (SDYS). SGBS is a condition characterized by pre- and postnatal overgrowth (gigantism) with visceral and skeletal anomalies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glypican proteoglycan 3; Glypican-3; GTR2-2; GTR22; heparan sulphate proteoglycan; Intestinal protein OCI-5; MXR7; secreted glypican-3
Gene Aliases: DGSX; GPC3; GTR2-2; MXR7; OCI-5; OCI5; SDYS; SGB; SGBS; SGBS1
UniProt ID: (Human) P51654
Entrez Gene ID: (Human) 2719
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support