Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 2
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration.
Product # 31140 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications.
Product # 31140 reacts with the heavy chain portion of human IgA, but not with the heavy chains of human IgG or IgM. The antibody does not react against the light chains common to most human immunoglobulins, or against non-immunoglobulin serum proteins. However, this antibody may cross-react with IgA from other species.
Store product at 4°C until opened. After opening, dilute only enough antibody for a single day's use. Store remainder at 4°C under sterile conditions.
Country of Origin: USA
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.