Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Description: This 1A8 monoclonal antibody reacts with human, mouse, and rat Grim-19. This 16 kDa protein is expressed in many human tissues, especially in the heart and skeletal muscle. Lower expression levels of Grim-19 can be observed in the liver, kidney, and brain. Originally described as a nuclear protein, reports indicate that Grim-19 is primarily localized to mitochondria as an essential component of complex I of the electron transport chain. Grim-19 expression can be induced by interferon-beta and all-trans-retinoic acid in tumor cell lines, leading to cell death. Finally, Grim-19 has been shown to bind the transactivation domain of STAT3 and inhibit its transcriptional activity. The molecular mechanisms of how Grim-19 regulates STAT3 within the cell remains under active investigation.
Applications Reported: This 1A8 antibody has been reported for use in immunoblotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and immunocytochemistry.
Applications Tested: This 1A8 antibody has been tested by immunoblotting of HeLa cell lysate. This can be used at less than or equal to 5 µg/mL. It is recommended that the antibody be titrated for optimal performance in the assay of interest.
Purity: Greater than 90%, as determined by SDS-PAGE.
Aggregation: Less than 10%, as determined by HPLC.
Filtration: 0.2 µm post-manufacturing filtered.
A novel gene associated with Retinoid-IFN-induced Mortality (GRIM) GRIM-19 gene was identified. Antisense expression of GRIM-19 confers a strong resistance against IFN/RA-induced death by reducing the intracellular levels of GRIM-19 protein. Overexpression of GRIM-19 enhances cell death in response to IFN/RA. GRIM-19 is primarily a nuclear protein whose expression is induced by the IFN/RA combination. These data indicate that GRIM-19 is a novel cell death-regulatory molecule.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell d; Cell death regulatory protein GRIM-19; cell death-regulatory protein GRIM19; CI-B16.6; complex I B16.6 subunit; Complex I-B16.6; FLJ58045; FLJ59191; gene associated with retinoic and IFN-induced mortality 19 protein; Gene associated with retinoic and interferon-induced mortality 19 protein; gene associated with retinoic-interferon-induced mortality 19 protein; genes associated with retinoid-IFN-induced mortality 19; GRIM-19; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13; NADH-ubiquinone oxidoreductase B16.6 subunit
Gene Aliases: 2700054G14Rik; AU022060; B16.6; CDA016; CGI-39; GRIM-19; GRIM19; NDUFA13
UniProt ID: (Human) Q9P0J0, (Mouse) Q9ERS2
Entrez Gene ID: (Rat) 100911483, (Human) 51079, (Mouse) 67184
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support