Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 9
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene encodes a member of the histone H2A family, and generates two transcripts through the use of the conserved stem-loop termination motif, and the polyA addition motif.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 11ac; Acetyl-Histone H2A.Z K4 K7 K11; Acetyl-Histone H2A.Z Lys4 Lys7 Lys11; H2A histone family, member V; H2A histone family, member Z; H2A.F/Z; H2A.Z variant histone 2; H2A.ZK4,7,11ac; H2A.ZK5; H2A.ZK5ac; H2A.ZLys4,7,11ac; H2A/z; H2AFZac; H2AZ histone; H2AZK11ac; H2AZK4,7,11ac; H2AZK7ac; H2AZLys4,7,11ac; histone H2A.F/Z; Histone H2A.V; Histone H2A.Z; purine-rich binding element protein B
Gene Aliases: H2A.z; H2A.Z-1; H2A.Z-2; H2A/z; H2AFV; H2AFZ; H2AV; H2AZ; H2AZ1; H2AZ2
UniProt ID: (Human) Q71UI9, (Human) P0C0S5
Entrez Gene ID: (Human) 94239, (Human) 3015
Molecular Function: histone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support