Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The enzyme encoded by the HAGH gene is classified as a thiolesterase and is responsible for the hydrolysis of S-lactoyl-glutathione to reduced glutathione and D-lactate. Three transcript variants encoding different isoforms have been found for this gene. Diseases associated with HAGH include Hydroxyacyl Glutathione Hydrolase Deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glx II; Glyoxalase II; glyoxylase 2; Hydroxyacylglutathione hydrolase, mitochondrial; hydroxyacylglutathione hydroxylase; Round spermatid protein RSP29
Gene Aliases: BC019817; Glo-2; GLO2; GLX2; GLXII; HAGH; HAGH1; RSP29
UniProt ID: (Human) Q16775, (Mouse) Q99KB8, (Rat) O35952
Entrez Gene ID: (Human) 3029, (Mouse) 14651, (Rat) 24439
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support