Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
HAND1 belongs to the basic helix-loop-helix family of transcription factors. This gene product is one of two closely related family members, the HAND proteins, which are asymmetrically expressed in the developing ventricular chambers and play an essential role in cardiac morphogenesis. Working in a complementary fashion, they function in the formation of the right ventricle and aortic arch arteries, implicating them as mediators of congenital heart disease. In addition, it has been suggested that this transcription factor may be required for early trophoblast differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHa27; Class A basic helix-loop-helix protein 27; eHAND; Extraembryonic tissues, heart, autonomic nervous system and neural crest derivatives-expressed protein 1; Heart- and neural crest derivatives-expressed protein 1
Gene Aliases: BHLHA27; EHAND; HAND1; Hxt; Thing1
UniProt ID: (Human) O96004
Entrez Gene ID: (Human) 9421
Molecular Function:
basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support