Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Has 2-hydroxyacid oxidase activity. Most active on the 2-carbon substrate glycolate, but is also active on 2-hydroxy fatty acids, with high activity towards 2-hydroxy palmitate and 2-hydroxy octanoate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (S)-2-hydroxy-acid oxidase; 2-Hydroxyacid oxidase 1; Glycolate oxidase; glycolate oxidase 1; Glyoxylate oxidase; GO; GOX1 antibody; HAOX1; HAOX1 antibody; hydroxyacid oxidase (glycolate oxidase) 1; MGC142225 antibody
Gene Aliases: GOX; GOX1; HAO1; HAOX1
UniProt ID: (Human) Q9UJM8
Entrez Gene ID: (Human) 54363
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support