Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSLVCLTDFQ AHAREQLSKS TRDFIEGGAD DSITRDDNIA AFKRIRLRPR YLRDVSEVDT RTTIQGEEIS APICIAPTGF HCLVWPDGEM STARAAQAAG ICYITSTFAS CSLEDIVIAA PEGLRWFQLY VHPDLQLNKQ LIQRVESLGF KALVITLDTP VCGNRRHDIR NQLRRNLTLT DLQSPKKGNA IPYFQMTPIS TSLCWNDLSW FQSITRLPII LKGILTKEDA ELAVKHNVQG IIVSNHGGRQ LDEVLASIDA LTEVVAAVKG KIEVYLDGGV RTGNDVLKAL ALGAKCIFLG RPILWGLACK GEHGVKEVLN ILTNEFHTSM ALTGCRSVAE INRNLVQFSR L
HAO2 gene ontology annotations related to this gene include fatty acid oxidation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (S)-2-hydroxy-acid oxidase, peroxisomal; 2-Hydroxyacid oxidase 2; Cell growth-inhibiting gene 16 protein; glycolate oxidase; HAOX2; hydroxyacid oxidase 2 (long chain); Long chain alpha-hydroxy acid oxidase; Long-chain L-2-hydroxy acid oxidase
Gene Aliases: GIG16; HAO2; HAOX2
UniProt ID: (Human) Q9NYQ3
Entrez Gene ID: (Human) 51179
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support