Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LREEASQLD TLEDEEQMLI LECVEQFSEA SQQMAELSEV LVLRLENYER QQQEVARLQA QVLKLQQRCR MYGAETEKLQ KQLASEKEIQ MQLQEESVWV GSQLQDLREK YMDCGGMLIE MQEEVKTLRQ QPPVSTGSAT HYPYSVPLET LPGFQETLAE ELRTSLRRMI SDPVYFMERN YEMPRGDTSS LRYDFRYSED REQVRGFEAE EGLMLAADIM RGEDFTPAEE FVPQEELGAA KKVPAEEGVM EEAELVSEET EGWEEVELEL DEATRMNVVT SALEASGLGP SHLDMNYVLQ QLANWQDAHY RRQLRWKMLQ KGECPHGALP AASRTSCRSS CR (331-671 aa encoded by BC156728)
Huntington's disease (HD), a neurodegenerative disorder characterized by loss of striatal neurons, is caused by an expansion of a polyglutamine tract in the HD protein huntingtin. HAP1 was initially identified through a two-hybrid library screening; the binding of HAP1 to huntingtin correlated with the expansion of the polyglutamine tract. HAP1 also interacts with two cytoskeletal proteins (dynactin and pericentriolar autoantigen protein 1), suggesting that HAP1 may play a role in vesicular trafficking or organelle transport. HAP1 is also involved with the huntingtin-enhanced BDNF transport along the cellular microtubles. Attenuation of this process led to the loss of neurotrophic support and neuronal toxicity, which suggests that loss of this function might contribute to pathogenesis. Several alternatively spliced isoforms have been described for HAP1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HAP-1; HAP-1 antibody; Huntingtin-associated protein 1; huntingtin-associated protein 2; Neuroan 1
Gene Aliases: HAP-1; HAP1; HAP2; hHLP1; HIP5; HLP; HLP1
UniProt ID: (Human) P54257, (Mouse) O35668
Entrez Gene ID: (Human) 9001, (Mouse) 15114
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support