Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MRPKRLGRCC AGSRLGPGDP AALTCAPSPS ASPAPEPSAQ PQARGTGQRV GSRATSGSQF LSEARTGARP ASEAGAKAGA RRPSAFSAIQ GDVRSMPDNS DAPWTRFVFQ GPFGSRATGR GTGKAAGIWK TPAAYVGRRP GVSGPERAAF IRELEEALCP NLPPPVKKIT QEDVKVMLYL LEELLPPVWE SVTYGMVLQR ERDLNTAARI GQSLVKQNSV LMEENSKLEA LLGSAKEEIL YLRHQVNLRD ELLQLYSDSD EEDEDEEEEE EEKEAEEEQE EEEAEEDLQC AHPCDAPKLI SQEALLHQHH CPQLEALQEK LRLLEEENHQ LREEASQLDT LEDEEQMLIL ECVEQFSEAS QQMAELSEVL VLRLENYERQ QQEVARLQAQ VLKLQQRCRM YGAETEKLQK QLASEKEIQM QLQEESVWVG SQLQDLREKY MDCGGMLIEM QEEVKTLRQQ PPVSTGSATH YPYSVPLETL PGFQETLAEE LRTSLRRMIS DPVYFMERNY EMPRGDTSSL RYDFRYSEDR EQVRGFEAEE GLMLAADIMR GEDFTPAEEF VPQEELGAAK KVPAEEGVME EAELVSEETE GWEEVELELD EATRMNVVTS ALEASGLGPS HLDMNYVLQQ LANWQDAHYR RQLRWKMLQK GECPHGALPA ASRTSCRSSC R
Huntington's disease (HD), a neurodegenerative disorder characterized by loss of striatal neurons, is caused by an expansion of a polyglutamine tract in the HD protein huntingtin. HAP1 was initially identified through a two-hybrid library screening; the binding of HAP1 to huntingtin correlated with the expansion of the polyglutamine tract. HAP1 also interacts with two cytoskeletal proteins (dynactin and pericentriolar autoantigen protein 1), suggesting that HAP1 may play a role in vesicular trafficking or organelle transport. HAP1 is also involved with the huntingtin-enhanced BDNF transport along the cellular microtubles. Attenuation of this process led to the loss of neurotrophic support and neuronal toxicity, which suggests that loss of this function might contribute to pathogenesis. Several alternatively spliced isoforms have been described for HAP1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HAP-1; HAP-1 antibody; Huntingtin-associated protein 1; huntingtin-associated protein 2; Neuroan 1
Gene Aliases: HAP1; HAP2; hHLP1; HIP5; HLP; HLP1
UniProt ID: (Human) P54257
Entrez Gene ID: (Human) 9001
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support