Product may be used with Western Blot (Cell lysate).
Immunogen sequence: QRGRKKVVHV LEGESGSVVV QTAPGQVVSH RGGTIVLPCR YHYEAAAHGH DGVRLKWTKV VDPLAFTDVF VALGPQHRAF GSYRGRAELQ GDGPGDASLV LRNVT
HAPLN4 binds to hyaluronic acid and may be involved in formation of the extracellular matrix.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain link protein 2; brain linked protein 2; Hyaluronan and proteoglycan link protein 4; link protein 4
Gene Aliases: 9330174O11; BRAL2; HAPLN4; KIAA1926; Lpr4
UniProt ID: (Human) Q86UW8, (Mouse) Q80WM4
Entrez Gene ID: (Human) 404037, (Mouse) 330790
Molecular Function:
extracellular matrix glycoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support