Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: QRGRKKVVHV LEGESGSVVV QTAPGQVVSH RGGTIVLPCR YHYEAAAHGH DGVRLKWTKV VDPLAFTDVF VALGPQHRAF GSYRGRAELQ GDGPGDASLV LRNVT
HAPLN4 binds to hyaluronic acid and may be involved in formation of the extracellular matrix.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain link protein 2; Hyaluronan and proteoglycan link protein 4
Gene Aliases: BRAL2; HAPLN4; KIAA1926
UniProt ID: (Human) Q86UW8
Entrez Gene ID: (Human) 404037
Molecular Function:
extracellular matrix glycoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support