Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a subunit of the augmin complex, which regulates centrosome and mitotic spindle integrity, and is necessary for the completion of cytokinesis. The encoded protein was identified by interaction with ubiquitin C-terminal hydrolase 37. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 26S proteasome-associated UCH37-interacting protein 1; HAUS augmin-like complex subunit 7; HAUS augmin-like complex, subunit 7; UCHL5 interacting protein; UCHL5-interacting protein; X-linked protein STS1769
Gene Aliases: HAUS7; UCHL5IP; UIP1
UniProt ID: (Human) Q99871
Entrez Gene ID: (Human) 55559
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support