Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Homeobox protein HB9 is expressed in lymphoid and pancreatic tissues. It is reported to be Putative transcription factor involved in pancreas development and function. Defects in this gene causes Currarino syndrome. The triad of a presacral tumor, sacral agenesis and anorectal malformation constitutes the Currarino syndrome which is caused by dorsal-ventral patterning defects during embryonic development. It is also reported that HB9 is strongly expressed in epidermal basal cells of the outer scale face in tarsometatarsal scale skin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeobox HB9; Homeobox protein HB9; HOX SCRA1; Motor neuron and pancreas homeobox protein 1
Gene Aliases: HB9; HLXB9; HOXHB9; MNX1; SCRA1
UniProt ID: (Human) P50219
Entrez Gene ID: (Human) 3110
Molecular Function:
helix-turn-helix transcription factor
homeodomain transcription factor
DNA-binding transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support