Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 8
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Hemoglobin subunit delta is a protein that in humans is encoded by the HBD gene. The delta (HBD) and beta (HBB) genes are normally expressed in the adult: two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin. Two alpha chains plus two delta chains constitute HbA-2, which with HbF comprises the remaining 3% of adult hemoglobin. Five beta-like globin genes are found within a 45 kb cluster on chromosome 11 in the following order: 5'-epsilon-Ggamma-Agamma-delta-beta-3'.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: delta globin; Delta-globin; delta-globin chain; Hemoglobin delta chain; Hemoglobin subunit delta; hemoglobin, delta
Gene Aliases: HBD
UniProt ID: (Human) P02042
Entrez Gene ID: (Human) 3045
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support