Product References
ZNF133 is a potent suppressor in breast carcinogenesis through dampening L1CAM, a driver for tumor progression.
Oncogene
Li L,Wang X,Hu K,Liu X,Qiu L,Bai C,Cui Y,Wang B,Wang Z,Wang H,Cheng R,Hua J,Hai L,Wang M,Liu M,Song Z,Xiao C,Li B
Thu Jun 01 00:00:00 EDT 2023
ZNF133 is a potent suppressor in breast carcinogenesis through dampening L1CAM, a driver for tumor progression.
Oncogene
Li L,Wang X,Hu K,Liu X,Qiu L,Bai C,Cui Y,Wang B,Wang Z,Wang H,Cheng R,Hua J,Hai L,Wang M,Liu M,Song Z,Xiao C,Li B
Thu Jun 01 00:00:00 EDT 2023
B-lymphoid tyrosine kinase-mediated FAM83A phosphorylation elevates pancreatic tumorigenesis through interacting with β-catenin.
Signal transduction and targeted therapy
Zhou C,Zhu X,Liu N,Dong X,Zhang X,Huang H,Tang Y,Liu S,Hu M,Wang M,Deng X,Li S,Zhang R,Huang Y,Lyu H,Xiao S,Luo S,Ali DW,Michalak M,Chen XZ,Wang Z,Tang J
Fri Feb 17 00:00:00 EST 2023
B-lymphoid tyrosine kinase-mediated FAM83A phosphorylation elevates pancreatic tumorigenesis through interacting with β-catenin.
Signal transduction and targeted therapy
Zhou C,Zhu X,Liu N,Dong X,Zhang X,Huang H,Tang Y,Liu S,Hu M,Wang M,Deng X,Li S,Zhang R,Huang Y,Lyu H,Xiao S,Luo S,Ali DW,Michalak M,Chen XZ,Wang Z,Tang J
Fri Feb 17 00:00:00 EST 2023
Transcription factor ETS proto-oncogene 1 contributes to neuropathic pain by regulating histone deacetylase 1 in primary afferent neurons.
Molecular pain
Zheng HL,Sun SY,Jin T,Zhang M,Zeng Y,Liu Q,Yang K,Wei R,Pan Z,Lin F
Sun Feb 12 00:00:00 EST 2023
Transcription factor ETS proto-oncogene 1 contributes to neuropathic pain by regulating histone deacetylase 1 in primary afferent neurons.
Molecular pain
Zheng HL,Sun SY,Jin T,Zhang M,Zeng Y,Liu Q,Yang K,Wei R,Pan Z,Lin F
Sun Feb 12 00:00:00 EST 2023
All-trans retinoic acid enhances the cytotoxic effect of decitabine on myelodysplastic syndromes and acute myeloid leukaemia by activating the RARα-Nrf2 complex.
British journal of cancer
Wang L,Zhang Q,Ye L,Ye X,Yang W,Zhang H,Zhou X,Ren Y,Ma L,Zhang X,Mei C,Xu G,Li K,Luo Y,Jiang L,Lin P,Zhu S,Lang W,Wang Y,Shen C,Han Y,Liu X,Yang H,Lu C,Sun J,Jin J,Tong H
Wed Feb 01 00:00:00 EST 2023
Peptidase Inhibitor 16 Attenuates Left Ventricular Injury and Remodeling After Myocardial Infarction by Inhibiting the HDAC1-Wnt3a-β-Catenin Signaling Axis.
Journal of the American Heart Association
Wang L,Du A,Lu Y,Zhao Y,Qiu M,Su Z,Shu H,Shen H,Sun W,Kong X
Tue May 16 00:00:00 EDT 2023
Combination with vorinostat enhances the antitumor activity of cisplatin in castration-resistant prostate cancer by inhibiting DNA damage repair pathway and detoxification of GSH.
The Prostate
Chang T,Lian Z,Ma S,Liang Z,Ma X,Wen X,Wang Y,Liu R
Sat Apr 01 00:00:00 EDT 2023
ARID1A Inactivation Increases Expression of circ0008399 and Promotes Cisplatin Resistance in Bladder Cancer.
Current medical science
Jiang YK,Shuai YJ,Ding HM,Zhang H,Huang C,Wang L,Sun JY,Wei WJ,Xiao XY,Jiang GS
Thu Jun 01 00:00:00 EDT 2023
ARID1A Inactivation Increases Expression of circ0008399 and Promotes Cisplatin Resistance in Bladder Cancer.
Current medical science
Jiang YK,Shuai YJ,Ding HM,Zhang H,Huang C,Wang L,Sun JY,Wei WJ,Xiao XY,Jiang GS
Thu Jun 01 00:00:00 EDT 2023
Hepatic RACK1 deficiency protects against fulminant hepatitis through myeloid-derived suppressor cells.
Theranostics
Liu G,Wang Q,Deng L,Huang X,Yang G,Cheng Q,Guo T,Guo L,Niu C,Yang X,Dong J,Zhang J
Thu Apr 21 00:00:00 EDT 2022
Hepatic RACK1 deficiency protects against fulminant hepatitis through myeloid-derived suppressor cells.
Theranostics
Liu G,Wang Q,Deng L,Huang X,Yang G,Cheng Q,Guo T,Guo L,Niu C,Yang X,Dong J,Zhang J
Thu Apr 21 00:00:00 EDT 2022
Hepatic RACK1 deficiency protects against fulminant hepatitis through myeloid-derived suppressor cells.
Theranostics
Liu G,Wang Q,Deng L,Huang X,Yang G,Cheng Q,Guo T,Guo L,Niu C,Yang X,Dong J,Zhang J
Thu Apr 21 00:00:00 EDT 2022
Hepatic RACK1 deficiency protects against fulminant hepatitis through myeloid-derived suppressor cells.
Theranostics
Liu G,Wang Q,Deng L,Huang X,Yang G,Cheng Q,Guo T,Guo L,Niu C,Yang X,Dong J,Zhang J
Thu Apr 21 00:00:00 EDT 2022
Histone deacetylase 1 and 3 inhibitors alleviate colon inflammation by inhibiting Th17 cell differentiation.
Journal of clinical laboratory analysis
Gu Z,Chen X,Zhu D,Wu S,Yu C
Sat Oct 01 00:00:00 EDT 2022
Gut microbiota shapes social dominance through modulating HDAC2 in the medial prefrontal cortex.
Cell reports
Wang T,Xu J,Xu Y,Xiao J,Bi N,Gu X,Wang HL
Tue Mar 08 00:00:00 EST 2022
Acetylation of p62 regulates base excision repair through interaction with APE1.
Cell reports
Li M,Xiong J,Yang L,Huang J,Zhang Y,Liu M,Wang L,Ji J,Zhao Y,Zhu WG,Luo J,Wang H
Tue Jul 19 00:00:00 EDT 2022
PCGF6 controls neuroectoderm specification of human pluripotent stem cells by activating SOX2 expression.
Nature communications
Lan X,Ding S,Zhang T,Yi Y,Li C,Jin W,Chen J,Liang K,Wang H,Jiang W
Sat Aug 06 00:00:00 EDT 2022
KTN1-AS1, a SOX2-mediated lncRNA, activates epithelial-mesenchymal transition process in esophageal squamous cell carcinoma.
Scientific reports
Chen L,Lu J,Xu T,Yan Z,Guo Y,Dong Z,Guo W
Wed Nov 23 00:00:00 EST 2022
Histone deacetylase inhibitors inhibit cervical cancer growth through Parkin acetylation-mediated mitophagy.
Acta pharmaceutica Sinica. B
Sun X,Shu Y,Ye G,Wu C,Xu M,Gao R,Huang D,Zhang J
Tue Feb 01 00:00:00 EST 2022
Post-translational control of beige fat biogenesis by PRDM16 stabilization.
Nature
Wang Q,Li H,Tajima K,Verkerke ARP,Taxin ZH,Hou Z,Cole JB,Li F,Wong J,Abe I,Pradhan RN,Yamamuro T,Yoneshiro T,Hirschhorn JN,Kajimura S
Thu Sep 01 00:00:00 EDT 2022
Disturbing NLRP3 acetylation and inflammasome assembly inhibits androgen receptor-promoted inflammatory responses and prostate cancer progression.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Zhao AN,Yang Z,Wang DD,Shi B,Zhang H,Bai Y,Yan BW,Zhang Y,Wen JK,Wang XL,Qu CB
Tue Nov 01 00:00:00 EDT 2022
miR-AB, a miRNA-based shRNA viral toolkit for multicolor-barcoded multiplex RNAi at a single-cell level.
EMBO reports
Wang D,Xiu J,Zhao J,Luo J
Tue Apr 05 00:00:00 EDT 2022
Damage of uremic myocardium by p-cresyl sulfate and the ameliorative effect of Klotho by regulating SIRT6 ubiquitination.
Toxicology letters
Chen C,Xie C,Xiong Y,Wu H,Wu L,Zhu J,Xing C,Mao H
Mon Aug 15 00:00:00 EDT 2022
The function of Sphingosine-1-phosphate receptor 2 (S1PR2) in maintaining intestinal barrier and inducing ulcerative colitis.
Bioengineered
Chen T,Gu K,Lin R,Liu Y,Shan Y
Sun May 01 00:00:00 EDT 2022
Targeting HNRNPM Inhibits Cancer Stemness and Enhances Antitumor Immunity in Wnt-activated Hepatocellular Carcinoma.
Cellular and molecular gastroenterology and hepatology
Zhu GQ,Wang Y,Wang B,Liu WR,Dong SS,Chen EB,Cai JL,Wan JL,Du JX,Song LN,Chen SP,Yu L,Zhou ZJ,Wang Z,Zhou J,Shi YH,Fan J,Dai Z
Mon Apr 18 00:00:00 EDT 2022
The histone modification reader ZCWPW1 promotes double-strand break repair by regulating cross-talk of histone modifications and chromatin accessibility at meiotic hotspots.
Genome biology
Yuan S,Huang T,Bao Z,Wang S,Wu X,Liu J,Liu H,Chen ZJ
Tue Sep 06 00:00:00 EDT 2022
The histone modification reader ZCWPW1 promotes double-strand break repair by regulating cross-talk of histone modifications and chromatin accessibility at meiotic hotspots.
Genome biology
Yuan S,Huang T,Bao Z,Wang S,Wu X,Liu J,Liu H,Chen ZJ
Tue Sep 06 00:00:00 EDT 2022
The histone modification reader ZCWPW1 promotes double-strand break repair by regulating cross-talk of histone modifications and chromatin accessibility at meiotic hotspots.
Genome biology
Yuan S,Huang T,Bao Z,Wang S,Wu X,Liu J,Liu H,Chen ZJ
Tue Sep 06 00:00:00 EDT 2022
The histone modification reader ZCWPW1 promotes double-strand break repair by regulating cross-talk of histone modifications and chromatin accessibility at meiotic hotspots.
Genome biology
Yuan S,Huang T,Bao Z,Wang S,Wu X,Liu J,Liu H,Chen ZJ
Tue Sep 06 00:00:00 EDT 2022
Sorafenib Attenuates Fibrotic Hepatic Injury Through Mediating Lysine Crotonylation.
Drug design, development and therapy
Chen XF,Ji S
Tue Jul 12 00:00:00 EDT 2022
PFKM inhibits doxorubicin-induced cardiotoxicity by enhancing oxidative phosphorylation and glycolysis.
Scientific reports
Zhou M,Sun X,Wang C,Wang F,Fang C,Hu Z
Fri Jul 08 00:00:00 EDT 2022
ZNF382 controls mouse neuropathic pain via silencer-based epigenetic inhibition of Cxcl13 in DRG neurons.
The Journal of experimental medicine
Ma L,Yu L,Jiang BC,Wang J,Guo X,Huang Y,Ren J,Sun N,Gao DS,Ding H,Lu J,Zhou H,Zou L,Gao Y,Wang L,Sun K,Ming Y,Meng Z,Tao YX,Yan M
Mon Dec 06 00:00:00 EST 2021
ZNF382 controls mouse neuropathic pain via silencer-based epigenetic inhibition of Cxcl13 in DRG neurons.
The Journal of experimental medicine
Ma L,Yu L,Jiang BC,Wang J,Guo X,Huang Y,Ren J,Sun N,Gao DS,Ding H,Lu J,Zhou H,Zou L,Gao Y,Wang L,Sun K,Ming Y,Meng Z,Tao YX,Yan M
Mon Dec 06 00:00:00 EST 2021
ZNF382 controls mouse neuropathic pain via silencer-based epigenetic inhibition of Cxcl13 in DRG neurons.
The Journal of experimental medicine
Ma L,Yu L,Jiang BC,Wang J,Guo X,Huang Y,Ren J,Sun N,Gao DS,Ding H,Lu J,Zhou H,Zou L,Gao Y,Wang L,Sun K,Ming Y,Meng Z,Tao YX,Yan M
Mon Dec 06 00:00:00 EST 2021
Epigenetic derepression converts PPARγ into a druggable target in triple-negative and endocrine-resistant breast cancers.
Cell death discovery
Loo SY,Syn NL,Koh AP,Teng JC,Deivasigamani A,Tan TZ,Thike AA,Vali S,Kapoor S,Wang X,Wang JW,Tan PH,Yip GW,Sethi G,Huang RY,Hui KM,Wang L,Goh BC,Kumar AP
Mon Sep 27 00:00:00 EDT 2021
The methyltransferase METTL3 negatively regulates nonalcoholic steatohepatitis (NASH) progression.
Nature communications
Li X,Yuan B,Lu M,Wang Y,Ding N,Liu C,Gao M,Yao Z,Zhang S,Zhao Y,Xie L,Chen Z
Fri Dec 10 00:00:00 EST 2021
LncRNA ZNFTR functions as an inhibitor in pancreatic cancer by modulating ATF3/ZNF24/VEGFA pathway.
Cell death & disease
Li W,Han S,Hu P,Chen D,Zeng Z,Hu Y,Xu F,Tang J,Wang F,Zhao Y,Huang M,Zhao G
Fri Sep 03 00:00:00 EDT 2021
LncRNA ZNFTR functions as an inhibitor in pancreatic cancer by modulating ATF3/ZNF24/VEGFA pathway.
Cell death & disease
Li W,Han S,Hu P,Chen D,Zeng Z,Hu Y,Xu F,Tang J,Wang F,Zhao Y,Huang M,Zhao G
Fri Sep 03 00:00:00 EDT 2021
MIER3 induces epithelial-mesenchymal transition and promotes breast cancer cell aggressiveness via forming a co-repressor complex with HDAC1/HDAC2/Snail.
Experimental cell research
Huang W,Chen J,Liu X,Liu X,Duan S,Chen L,Liu X,Lan J,Zou Y,Guo D,Zhou J
Wed Sep 01 00:00:00 EDT 2021
MIER3 induces epithelial-mesenchymal transition and promotes breast cancer cell aggressiveness via forming a co-repressor complex with HDAC1/HDAC2/Snail.
Experimental cell research
Huang W,Chen J,Liu X,Liu X,Duan S,Chen L,Liu X,Lan J,Zou Y,Guo D,Zhou J
Wed Sep 01 00:00:00 EDT 2021
Activation of TAF9 via Danshensu-Induced Upregulation of HDAC1 Expression Alleviates Non-alcoholic Fatty Liver Disease.
Frontiers in pharmacology
Wang R,Wang Z,Sun R,Fu R,Sun Y,Zhu M,Geng Y,Gao D,Tian X,Zhao Y,Yao J
Tue Dec 21 00:00:00 EST 2021
Activation of TAF9 via Danshensu-Induced Upregulation of HDAC1 Expression Alleviates Non-alcoholic Fatty Liver Disease.
Frontiers in pharmacology
Wang R,Wang Z,Sun R,Fu R,Sun Y,Zhu M,Geng Y,Gao D,Tian X,Zhao Y,Yao J
Tue Dec 21 00:00:00 EST 2021
Activation of TAF9 via Danshensu-Induced Upregulation of HDAC1 Expression Alleviates Non-alcoholic Fatty Liver Disease.
Frontiers in pharmacology
Wang R,Wang Z,Sun R,Fu R,Sun Y,Zhu M,Geng Y,Gao D,Tian X,Zhao Y,Yao J
Tue Dec 21 00:00:00 EST 2021
Activation of TAF9 via Danshensu-Induced Upregulation of HDAC1 Expression Alleviates Non-alcoholic Fatty Liver Disease.
Frontiers in pharmacology
Wang R,Wang Z,Sun R,Fu R,Sun Y,Zhu M,Geng Y,Gao D,Tian X,Zhao Y,Yao J
Tue Dec 21 00:00:00 EST 2021
GCH1 induces immunosuppression through metabolic reprogramming and IDO1 upregulation in triple-negative breast cancer.
Journal for immunotherapy of cancer
Wei JL,Wu SY,Yang YS,Xiao Y,Jin X,Xu XE,Hu X,Li DQ,Jiang YZ,Shao ZM
Thu Jul 01 00:00:00 EDT 2021
PWWP2B Fine-Tunes Adipose Thermogenesis by Stabilizing HDACs in a NuRD Subcomplex.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Yan L,Jin W,Zhao Q,Cui X,Shi T,Xu Y,Li F,Jin W,Zhang Z,Zhang Z,Tang QQ,Pan D
Sun Aug 01 00:00:00 EDT 2021
PWWP2B Fine-Tunes Adipose Thermogenesis by Stabilizing HDACs in a NuRD Subcomplex.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Yan L,Jin W,Zhao Q,Cui X,Shi T,Xu Y,Li F,Jin W,Zhang Z,Zhang Z,Tang QQ,Pan D
Sun Aug 01 00:00:00 EDT 2021
Positive Reciprocal Feedback of lncRNA ZEB1-AS1 and HIF-1α Contributes to Hypoxia-Promoted Tumorigenesis and Metastasis of Pancreatic Cancer.
Frontiers in oncology
Jin Y,Zhang Z,Yu Q,Zeng Z,Song H,Huang X,Kong Q,Hu H,Xia Y
Fri Apr 29 00:00:00 EDT 2022
Hepatic COX-2 expression protects mice from an alcohol-high fat diet-induced metabolic disorder by involving protein acetylation related energy metabolism.
Alcohol (Fayetteville, N.Y.)
Chen M,Sun X,Wei W,Cucarella C,Martín-Sanz P,Casado M,Pi L,Ren B,Cao Q
Sat May 01 00:00:00 EDT 2021
Low levels of AMPK promote epithelial-mesenchymal transition in lung cancer primarily through HDAC4- and HDAC5-mediated metabolic reprogramming.
Journal of cellular and molecular medicine
Feng S,Zhang L,Liu X,Li G,Zhang B,Wang Z,Zhang H,Ma H
Wed Jul 01 00:00:00 EDT 2020
USP38 regulates the stemness and chemoresistance of human colorectal cancer via regulation of HDAC3.
Oncogenesis
Zhan W,Liao X,Liu J,Tian T,Yu L,Li R
Wed May 13 00:00:00 EDT 2020
USP38 regulates the stemness and chemoresistance of human colorectal cancer via regulation of HDAC3.
Oncogenesis
Zhan W,Liao X,Liu J,Tian T,Yu L,Li R
Wed May 13 00:00:00 EDT 2020
Signatures of host-pathogen evolutionary conflict reveal MISTR-A conserved MItochondrial STress Response network.
PLoS biology
Sorouri M,Chang T,Jesudhasan P,Pinkham C,Elde NC,Hancks DC
Tue Dec 01 00:00:00 EST 2020
Increased NRSF/REST in anterior cingulate cortex contributes to diabetes-related neuropathic pain.
Biochemical and biophysical research communications
Xiao-Die X,Xiao-Hong W,Cheng-Feng H,Zhong-Yu Y,Jian-Tao W,Hou-Guang Z,Jing-Chun G
Tue Jun 30 00:00:00 EDT 2020
HDAC1 inhibition ameliorates TDP-43-induced cell death in vitro and in vivo.
Cell death & disease
Sanna S,Esposito S,Masala A,Sini P,Nieddu G,Galioto M,Fais M,Iaccarino C,Cestra G,Crosio C
Thu May 14 00:00:00 EDT 2020
HDAC1 inhibition ameliorates TDP-43-induced cell death in vitro and in vivo.
Cell death & disease
Sanna S,Esposito S,Masala A,Sini P,Nieddu G,Galioto M,Fais M,Iaccarino C,Cestra G,Crosio C
Thu May 14 00:00:00 EDT 2020
HDAC1 inhibition ameliorates TDP-43-induced cell death in vitro and in vivo.
Cell death & disease
Sanna S,Esposito S,Masala A,Sini P,Nieddu G,Galioto M,Fais M,Iaccarino C,Cestra G,Crosio C
Thu May 14 00:00:00 EDT 2020
HDAC1 inhibition ameliorates TDP-43-induced cell death in vitro and in vivo.
Cell death & disease
Sanna S,Esposito S,Masala A,Sini P,Nieddu G,Galioto M,Fais M,Iaccarino C,Cestra G,Crosio C
Thu May 14 00:00:00 EDT 2020
Crosstalk between cardiomyocytes and noncardiomyocytes is essential to prevent cardiomyocyte apoptosis induced by proteasome inhibition.
Cell death & disease
Guo F,Zhang CC,Yin XH,Li T,Fang CH,He XB
Sat Sep 19 00:00:00 EDT 2020
The RNA exosome complex degrades expanded hexanucleotide repeat RNA in C9orf72 FTLD/ALS.
The EMBO journal
Kawabe Y,Mori K,Yamashita T,Gotoh S,Ikeda M
Thu Oct 01 00:00:00 EDT 2020
EGFR-vIII downregulated H2AZK4/7AC though the PI3K/AKT-HDAC2 axis to regulate cell cycle progression.
Clinical and translational medicine
Zhao H,Wang Y,Yang C,Zhou J,Wang L,Yi K,Li Y,Wang Q,Shi J,Kang C,Zeng L
Tue Jan 28 00:00:00 EST 2020
EGFR-vIII downregulated H2AZK4/7AC though the PI3K/AKT-HDAC2 axis to regulate cell cycle progression.
Clinical and translational medicine
Zhao H,Wang Y,Yang C,Zhou J,Wang L,Yi K,Li Y,Wang Q,Shi J,Kang C,Zeng L
Tue Jan 28 00:00:00 EST 2020
Aberrant HDAC3 expression correlates with brain metastasis in breast cancer patients.
Thoracic cancer
Ma L,Qi L,Li S,Yin Q,Liu J,Wang J,She C,Li P,Liu Q,Wang X,Li W
Tue Sep 01 00:00:00 EDT 2020
Acetaldehyde dehydrogenase 2 interactions with LDLR and AMPK regulate foam cell formation.
The Journal of clinical investigation
Zhong S,Li L,Zhang YL,Zhang L,Lu J,Guo S,Liang N,Ge J,Zhu M,Tao Y,Wu YC,Yin H
Wed Jan 02 00:00:00 EST 2019
Acetaldehyde dehydrogenase 2 interactions with LDLR and AMPK regulate foam cell formation.
The Journal of clinical investigation
Zhong S,Li L,Zhang YL,Zhang L,Lu J,Guo S,Liang N,Ge J,Zhu M,Tao Y,Wu YC,Yin H
Wed Jan 02 00:00:00 EST 2019
SREBP1, targeted by miR-18a-5p, modulates epithelial-mesenchymal transition in breast cancer via forming a co-repressor complex with Snail and HDAC1/2.
Cell death and differentiation
Zhang N,Zhang H,Liu Y,Su P,Zhang J,Wang X,Sun M,Chen B,Zhao W,Wang L,Wang H,Moran MS,Haffty BG,Yang Q
Wed May 01 00:00:00 EDT 2019
SREBP1, targeted by miR-18a-5p, modulates epithelial-mesenchymal transition in breast cancer via forming a co-repressor complex with Snail and HDAC1/2.
Cell death and differentiation
Zhang N,Zhang H,Liu Y,Su P,Zhang J,Wang X,Sun M,Chen B,Zhao W,Wang L,Wang H,Moran MS,Haffty BG,Yang Q
Wed May 01 00:00:00 EDT 2019
Histone deacetylase-1 as a prognostic factor and mediator of gastric cancer progression by enhancing glycolysis.
Human pathology
Jiang Z,Yang H,Zhang X,Wang Z,Rong R,Wang X
Fri Mar 01 00:00:00 EST 2019
Histone deacetylase-1 as a prognostic factor and mediator of gastric cancer progression by enhancing glycolysis.
Human pathology
Jiang Z,Yang H,Zhang X,Wang Z,Rong R,Wang X
Fri Mar 01 00:00:00 EST 2019
Histone deacetylase-1 as a prognostic factor and mediator of gastric cancer progression by enhancing glycolysis.
Human pathology
Jiang Z,Yang H,Zhang X,Wang Z,Rong R,Wang X
Fri Mar 01 00:00:00 EST 2019
Histone deacetylase-1 as a prognostic factor and mediator of gastric cancer progression by enhancing glycolysis.
Human pathology
Jiang Z,Yang H,Zhang X,Wang Z,Rong R,Wang X
Fri Mar 01 00:00:00 EST 2019
Purification and enzymatic assay of class I histone deacetylase enzymes.
Methods in enzymology
Adams MK,Banks CAS,Miah S,Killer M,Washburn MP
Tue Jun 02 00:00:00 EDT 2020
Heartland virus antagonizes type I and III interferon antiviral signaling by inhibiting phosphorylation and nuclear translocation of STAT2 and STAT1.
The Journal of biological chemistry
Feng K,Deng F,Hu Z,Wang H,Ning YJ
Fri Jun 14 00:00:00 EDT 2019
Hypoxia-induced feedback of HIF-1α and lncRNA-CF129 contributes to pancreatic cancer progression through stabilization of p53 protein.
Theranostics
Liu M,Zhong J,Zeng Z,Huang K,Ye Z,Deng S,Chen H,Xu F,Li Q,Zhao G
Tue Aug 25 00:00:00 EDT 2020
Reversal of increased mammary tumorigenesis by valproic acid and hydralazine in offspring of dams fed high fat diet during pregnancy.
Scientific reports
Andrade FO,Nguyen NM,Warri A,Hilakivi-Clarke L
Mon Dec 30 00:00:00 EST 2019
Disruption of Planar Cell Polarity Pathway Attributable to Valproic Acid-Induced Congenital Heart Disease through Hdac3 Participation in Mice.
Chinese medical journal
Duan HY,Zhou KY,Wang T,Zhang Y,Li YF,Hua YM,Wang C
Wed Sep 05 00:00:00 EDT 2018
Disruption of Planar Cell Polarity Pathway Attributable to Valproic Acid-Induced Congenital Heart Disease through Hdac3 Participation in Mice.
Chinese medical journal
Duan HY,Zhou KY,Wang T,Zhang Y,Li YF,Hua YM,Wang C
Wed Sep 05 00:00:00 EDT 2018
Disruption of Planar Cell Polarity Pathway Attributable to Valproic Acid-Induced Congenital Heart Disease through Hdac3 Participation in Mice.
Chinese medical journal
Duan HY,Zhou KY,Wang T,Zhang Y,Li YF,Hua YM,Wang C
Wed Sep 05 00:00:00 EDT 2018
ABIN1 inhibits HDAC1 ubiquitination and protects it from both proteasome- and lysozyme-dependent degradation.
Journal of cellular biochemistry
Ma Y,Yuan S,Tian X,Lin S,Wei S,Hu T,Chen S,Li X,Chen S,Wu D,Wang M,Guo D
Sun Apr 01 00:00:00 EDT 2018
ABIN1 inhibits HDAC1 ubiquitination and protects it from both proteasome- and lysozyme-dependent degradation.
Journal of cellular biochemistry
Ma Y,Yuan S,Tian X,Lin S,Wei S,Hu T,Chen S,Li X,Chen S,Wu D,Wang M,Guo D
Sun Apr 01 00:00:00 EDT 2018
ABIN1 inhibits HDAC1 ubiquitination and protects it from both proteasome- and lysozyme-dependent degradation.
Journal of cellular biochemistry
Ma Y,Yuan S,Tian X,Lin S,Wei S,Hu T,Chen S,Li X,Chen S,Wu D,Wang M,Guo D
Sun Apr 01 00:00:00 EDT 2018
ABIN1 inhibits HDAC1 ubiquitination and protects it from both proteasome- and lysozyme-dependent degradation.
Journal of cellular biochemistry
Ma Y,Yuan S,Tian X,Lin S,Wei S,Hu T,Chen S,Li X,Chen S,Wu D,Wang M,Guo D
Sun Apr 01 00:00:00 EDT 2018
Impaired FGF10 Signaling and Epithelial Development in Experimental Lung Hypoplasia With Esophageal Atresia.
Frontiers in pediatrics
Wang J,Liu H,Gao L,Liu X
Sun Nov 12 00:00:00 EST 2023
Impaired FGF10 Signaling and Epithelial Development in Experimental Lung Hypoplasia With Esophageal Atresia.
Frontiers in pediatrics
Wang J,Liu H,Gao L,Liu X
Sun Nov 12 00:00:00 EST 2023
Impaired FGF10 Signaling and Epithelial Development in Experimental Lung Hypoplasia With Esophageal Atresia.
Frontiers in pediatrics
Wang J,Liu H,Gao L,Liu X
Sun Nov 12 00:00:00 EST 2023
Impaired FGF10 Signaling and Epithelial Development in Experimental Lung Hypoplasia With Esophageal Atresia.
Frontiers in pediatrics
Wang J,Liu H,Gao L,Liu X
Sun Nov 12 00:00:00 EST 2023
The HDAC Inhibitor Quisinostat (JNJ-26481585) Supresses Hepatocellular Carcinoma alone and Synergistically in Combination with Sorafenib by G0/G1 phase arrest and Apoptosis induction.
International journal of biological sciences
He B,Dai L,Zhang X,Chen D,Wu J,Feng X,Zhang Y,Xie H,Zhou L,Wu J,Zheng S
Mon Sep 09 00:00:00 EDT 2019
The HDAC Inhibitor Quisinostat (JNJ-26481585) Supresses Hepatocellular Carcinoma alone and Synergistically in Combination with Sorafenib by G0/G1 phase arrest and Apoptosis induction.
International journal of biological sciences
He B,Dai L,Zhang X,Chen D,Wu J,Feng X,Zhang Y,Xie H,Zhou L,Wu J,Zheng S
Mon Sep 09 00:00:00 EDT 2019
Differential HDAC1/2 network analysis reveals a role for prefoldin/CCT in HDAC1/2 complex assembly.
Scientific reports
Banks CAS,Miah S,Adams MK,Eubanks CG,Thornton JL,Florens L,Washburn MP
Wed Sep 12 00:00:00 EDT 2018
Differential HDAC1/2 network analysis reveals a role for prefoldin/CCT in HDAC1/2 complex assembly.
Scientific reports
Banks CAS,Miah S,Adams MK,Eubanks CG,Thornton JL,Florens L,Washburn MP
Wed Sep 12 00:00:00 EDT 2018
The roles of SMYD4 in epigenetic regulation of cardiac development in zebrafish.
PLoS genetics
Xiao D,Wang H,Hao L,Guo X,Ma X,Qian Y,Chen H,Ma J,Zhang J,Sheng W,Shou W,Huang G,Ma D
Wed Aug 01 00:00:00 EDT 2018
The roles of SMYD4 in epigenetic regulation of cardiac development in zebrafish.
PLoS genetics
Xiao D,Wang H,Hao L,Guo X,Ma X,Qian Y,Chen H,Ma J,Zhang J,Sheng W,Shou W,Huang G,Ma D
Wed Aug 01 00:00:00 EDT 2018
The roles of SMYD4 in epigenetic regulation of cardiac development in zebrafish.
PLoS genetics
Xiao D,Wang H,Hao L,Guo X,Ma X,Qian Y,Chen H,Ma J,Zhang J,Sheng W,Shou W,Huang G,Ma D
Wed Aug 01 00:00:00 EDT 2018
The roles of SMYD4 in epigenetic regulation of cardiac development in zebrafish.
PLoS genetics
Xiao D,Wang H,Hao L,Guo X,Ma X,Qian Y,Chen H,Ma J,Zhang J,Sheng W,Shou W,Huang G,Ma D
Wed Aug 01 00:00:00 EDT 2018
Toxoplasma Effector TgIST Targets Host IDO1 to Antagonize the IFN-γ-Induced Anti-parasitic Response in Human Cells.
Frontiers in immunology
Bando H,Sakaguchi N,Lee Y,Pradipta A,Ma JS,Tanaka S,Lai DH,Liu J,Lun ZR,Nishikawa Y,Sasai M,Yamamoto M
Mon Sep 30 00:00:00 EDT 2019
Toxoplasma Effector TgIST Targets Host IDO1 to Antagonize the IFN-γ-Induced Anti-parasitic Response in Human Cells.
Frontiers in immunology
Bando H,Sakaguchi N,Lee Y,Pradipta A,Ma JS,Tanaka S,Lai DH,Liu J,Lun ZR,Nishikawa Y,Sasai M,Yamamoto M
Mon Sep 30 00:00:00 EDT 2019
Regulatory Roles of Histone Deacetylases 1 and 2 in Pb-induced Neurotoxicity.
Toxicological sciences : an official journal of the Society of Toxicology
Wu Y,Xu Y,Huang X,Ye D,Han M,Wang HL
Sun Apr 01 00:00:00 EDT 2018
Regulatory Roles of Histone Deacetylases 1 and 2 in Pb-induced Neurotoxicity.
Toxicological sciences : an official journal of the Society of Toxicology
Wu Y,Xu Y,Huang X,Ye D,Han M,Wang HL
Sun Apr 01 00:00:00 EDT 2018
Reciprocal loop of hypoxia-inducible factor-1α (HIF-1α) and metastasis-associated protein 2 (MTA2) contributes to the progression of pancreatic carcinoma by suppressing E-cadherin transcription.
The Journal of pathology
Zhu S,Deng S,He C,Liu M,Chen H,Zeng Z,Zhong J,Ye Z,Deng S,Wu H,Wang C,Zhao G
Sun Jul 01 00:00:00 EDT 2018
Effect of Histone Deacetylase Inhibition on the Expression of Multidrug Resistance-associated Protein 2 in a Human Placental Trophoblast Cell Line.
Chinese medical journal
Duan HY,Ma D,Zhou KY,Wang T,Zhang Y,Li YF,Wu JL,Hua YM,Wang C
Mon Jun 05 00:00:00 EDT 2017
Effect of Histone Deacetylase Inhibition on the Expression of Multidrug Resistance-associated Protein 2 in a Human Placental Trophoblast Cell Line.
Chinese medical journal
Duan HY,Ma D,Zhou KY,Wang T,Zhang Y,Li YF,Wu JL,Hua YM,Wang C
Mon Jun 05 00:00:00 EDT 2017
The effect of histone deacetylase inhibition on the expression of P-glycoprotein in human placental trophoblast cell lines.
Placenta
Duan H,Wang C,Zhou K,Wang T,Li Y,Qiu D,Li Q,Zhang Y,Hua Y
Sun Jan 01 00:00:00 EST 2017
The effect of histone deacetylase inhibition on the expression of P-glycoprotein in human placental trophoblast cell lines.
Placenta
Duan H,Wang C,Zhou K,Wang T,Li Y,Qiu D,Li Q,Zhang Y,Hua Y
Sun Jan 01 00:00:00 EST 2017
HDAC2 was involved in placental P-glycoprotein regulation both in vitro and vivo.
Placenta
Duan H,Zhou K,Zhang Y,Yue P,Wang T,Li Y,Qiu D,Wu J,Hua Y,Wang C
Sun Oct 01 00:00:00 EDT 2017
miR-155 downregulates ErbB2 and suppresses ErbB2-induced malignant transformation of breast epithelial cells.
Oncogene
He XH,Zhu W,Yuan P,Jiang S,Li D,Zhang HW,Liu MF
Thu Nov 17 00:00:00 EST 2016
miR-155 downregulates ErbB2 and suppresses ErbB2-induced malignant transformation of breast epithelial cells.
Oncogene
He XH,Zhu W,Yuan P,Jiang S,Li D,Zhang HW,Liu MF
Thu Nov 17 00:00:00 EST 2016
Capsaicin reactivates hMOF in gastric cancer cells and induces cell growth inhibition.
Cancer biology & therapy
Wang F,Zhao J,Liu D,Zhao T,Lu Z,Zhu L,Cao L,Yang J,Jin J,Cai Y
Tue Nov 01 00:00:00 EDT 2016
Capsaicin reactivates hMOF in gastric cancer cells and induces cell growth inhibition.
Cancer biology & therapy
Wang F,Zhao J,Liu D,Zhao T,Lu Z,Zhu L,Cao L,Yang J,Jin J,Cai Y
Tue Nov 01 00:00:00 EDT 2016
The long non-coding RNA TUG1 indicates a poor prognosis for colorectal cancer and promotes metastasis by affecting epithelial-mesenchymal transition.
Journal of translational medicine
Sun J,Ding C,Yang Z,Liu T,Zhang X,Zhao C,Wang J
Mon Feb 08 00:00:00 EST 2016
The long non-coding RNA TUG1 indicates a poor prognosis for colorectal cancer and promotes metastasis by affecting epithelial-mesenchymal transition.
Journal of translational medicine
Sun J,Ding C,Yang Z,Liu T,Zhang X,Zhao C,Wang J
Mon Feb 08 00:00:00 EST 2016
Identification of NOD2 as a novel target of RNA-binding protein HuR: evidence from NADPH oxidase-mediated HuR signaling in diabetic nephropathy.
Free radical biology & medicine
Shang J,Wan Q,Wang X,Duan Y,Wang Z,Wei X,Zhang Y,Wang H,Wang R,Yi F
Sun Feb 01 00:00:00 EST 2015
Identification of NOD2 as a novel target of RNA-binding protein HuR: evidence from NADPH oxidase-mediated HuR signaling in diabetic nephropathy.
Free radical biology & medicine
Shang J,Wan Q,Wang X,Duan Y,Wang Z,Wei X,Zhang Y,Wang H,Wang R,Yi F
Sun Feb 01 00:00:00 EST 2015
Expression of hMOF in different ovarian tissues and its effects on ovarian cancer prognosis.
Oncology reports
Cai M,Hu Z,Liu J,Gao J,Tan M,Zhang D,Zhu L,Liu S,Hou R,Lin B
Sun Feb 01 00:00:00 EST 2015
Expression of hMOF, but not HDAC4, is responsible for the global histone H4K16 acetylation in gastric carcinoma.
International journal of oncology
Zhu L,Yang J,Zhao L,Yu X,Wang L,Wang F,Cai Y,Jin J
Tue Jan 26 00:00:00 EST 2016
Specific immunotherapy in combination with Clostridium butyricum inhibits allergic inflammation in the mouse intestine.
Scientific reports
Shi Y,Xu LZ,Peng K,Wu W,Wu R,Liu ZQ,Yang G,Geng XR,Liu J,Liu ZG,Liu Z,Yang PC
Wed Dec 02 00:00:00 EST 2015
Specific immunotherapy in combination with Clostridium butyricum inhibits allergic inflammation in the mouse intestine.
Scientific reports
Shi Y,Xu LZ,Peng K,Wu W,Wu R,Liu ZQ,Yang G,Geng XR,Liu J,Liu ZG,Liu Z,Yang PC
Wed Dec 02 00:00:00 EST 2015
Connexin32 regulates hepatoma cell metastasis and proliferation via the p53 and Akt pathways.
Oncotarget
Zhao B,Zhao W,Wang Y,Xu Y,Xu J,Tang K,Zhang S,Yin Z,Wu Q,Wang X
Thu Apr 30 00:00:00 EDT 2015
Valproic acid inhibits tumor angiogenesis in mice transplanted with Kasumi‑1 leukemia cells.
Molecular medicine reports
Zhang ZH,Hao CL,Liu P,Tian X,Wang LH,Zhao L,Zhu CM
Sat Feb 01 00:00:00 EST 2014
Valproic acid inhibits tumor angiogenesis in mice transplanted with Kasumi‑1 leukemia cells.
Molecular medicine reports
Zhang ZH,Hao CL,Liu P,Tian X,Wang LH,Zhao L,Zhu CM
Sat Feb 01 00:00:00 EST 2014
Histone cross-talk connects protein phosphatase 1α (PP1α) and histone deacetylase (HDAC) pathways to regulate the functional transition of bromodomain-containing 4 (BRD4) for inducible gene expression.
The Journal of biological chemistry
Hu X,Lu X,Liu R,Ai N,Cao Z,Li Y,Liu J,Yu B,Liu K,Wang H,Zhou C,Wang Y,Han A,Ding F,Chen R
Fri Aug 15 00:00:00 EDT 2014
Histone cross-talk connects protein phosphatase 1α (PP1α) and histone deacetylase (HDAC) pathways to regulate the functional transition of bromodomain-containing 4 (BRD4) for inducible gene expression.
The Journal of biological chemistry
Hu X,Lu X,Liu R,Ai N,Cao Z,Li Y,Liu J,Yu B,Liu K,Wang H,Zhou C,Wang Y,Han A,Ding F,Chen R
Fri Aug 15 00:00:00 EDT 2014
PROX1 promotes hepatocellular carcinoma metastasis by way of up-regulating hypoxia-inducible factor 1α expression and protein stability.
Hepatology (Baltimore, Md.)
Liu Y,Zhang JB,Qin Y,Wang W,Wei L,Teng Y,Guo L,Zhang B,Lin Z,Liu J,Ren ZG,Ye QH,Xie Y
Thu Aug 01 00:00:00 EDT 2013
PROX1 promotes hepatocellular carcinoma metastasis by way of up-regulating hypoxia-inducible factor 1α expression and protein stability.
Hepatology (Baltimore, Md.)
Liu Y,Zhang JB,Qin Y,Wang W,Wei L,Teng Y,Guo L,Zhang B,Lin Z,Liu J,Ren ZG,Ye QH,Xie Y
Thu Aug 01 00:00:00 EDT 2013
PROX1 promotes hepatocellular carcinoma metastasis by way of up-regulating hypoxia-inducible factor 1α expression and protein stability.
Hepatology (Baltimore, Md.)
Liu Y,Zhang JB,Qin Y,Wang W,Wei L,Teng Y,Guo L,Zhang B,Lin Z,Liu J,Ren ZG,Ye QH,Xie Y
Thu Aug 01 00:00:00 EDT 2013
PROX1 promotes hepatocellular carcinoma metastasis by way of up-regulating hypoxia-inducible factor 1α expression and protein stability.
Hepatology (Baltimore, Md.)
Liu Y,Zhang JB,Qin Y,Wang W,Wei L,Teng Y,Guo L,Zhang B,Lin Z,Liu J,Ren ZG,Ye QH,Xie Y
Thu Aug 01 00:00:00 EDT 2013
FLASH knockdown sensitizes cells to Fas-mediated apoptosis via down-regulation of the anti-apoptotic proteins, MCL-1 and Cflip short.
PloS one
Chen S,Evans HG,Evans DR
Mon Aug 20 00:00:00 EDT 2012
FLASH knockdown sensitizes cells to Fas-mediated apoptosis via down-regulation of the anti-apoptotic proteins, MCL-1 and Cflip short.
PloS one
Chen S,Evans HG,Evans DR
Mon Aug 20 00:00:00 EDT 2012
Modulation of Tcf3 repressor complex composition regulates cdx4 expression in zebrafish.
The EMBO journal
Ro H,Dawid IB
Fri Jun 10 00:00:00 EDT 2011
Modulation of Tcf3 repressor complex composition regulates cdx4 expression in zebrafish.
The EMBO journal
Ro H,Dawid IB
Fri Jun 10 00:00:00 EDT 2011