Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to HDAC6. It does not cross react with other HDACs.
Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Plays a central role in microtubule-dependent cell motility via deacetylation of tubulin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ16239; HD6; HDA6; histone deacetylase 5; Histone deacetylase 6; Histone deacetylase mHDA2; Protein deacetylase HDAC6; protein phosphatase 1, regulatory subunit 90; scurfy candidate 6; Tubulin-lysine deacetylase HDAC6
Gene Aliases: CPBHM; HD6; Hdac5; HDAC6; JM21; KIAA0901; mHDA2; PPP1R90; Sfc6
UniProt ID: (Human) Q9UBN7, (Mouse) B1AUA6
Entrez Gene ID: (Human) 10013, (Rat) 84581, (Mouse) 15185
Molecular Function:
histone modifying enzyme
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support