Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MARPRPREYK AGDLVFAKMK GYPHWPARID ELPEGAVKPP ANKYPIFFFG THETAFLGPK DLFPYKEYKD KFGKSNKRKG FNEGLWEIEN NPGVKFTGYQ AIQQQSSSET EGEGGNTADA SSEEEGDRVE EDGKGKRKNE KAGSKRKKSY TSKKSSKQSR KSPGDEDDKD CKEEENKSSS EGGDAGNDTR NTTSDLQKTS EGT
HDGFRP3 enhances DNA synthesis and may play a role in cell proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HDGF-2; Hepatoma-derived growth factor 2; Hepatoma-derived growth factor-related protein 3; HRP-3
Gene Aliases: CGI-142; HDGF-2; HDGF2; HDGFL3; HDGFRP3; HRP-3
UniProt ID: (Human) Q9Y3E1
Entrez Gene ID: (Human) 50810
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support