Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1. 6 kb and a protein product with a molecular weight of about 35 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DnaJ (Hsp40) homolog, subfamily A, member 1; DnaJ homolog subfamily A member 1; DnaJ protein homolog 2; h HSJ-2; hDj-2; Heat shock 40 kDa protein 4; Heat shock protein J2; heat shock protein, DNAJ-like 2; HSDJ; HSJ-2; Human DnaJ protein 2; neural precursor cell expressed, developmentally down-regulated 7
Gene Aliases: DJ-2; DjA1; DNAJ2; DNAJA1; hDJ-2; HDJ2; HSDJ; HSJ-2; HSJ2; HSPF4; NEDD7
UniProt ID: (Human) P31689
Entrez Gene ID: (Human) 3301
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support