Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NLQWHAGRT AAAIRTAAVS CLWALTSSEV LSAEQIRDVQ ETLMPQVLTT LEEDSKMTRL ISCRIINTFL KTSGGMTDPE KLIRIYPELL KRLDDVSNDV RMAAASTLVT WLQCVKGANA KSYYQSSVQY LYRELLVHLD DPERAIQDAI (661-809 aa encoded by BC047240)
The protein encoded by this gene is essential for the preassembly or stability of axonemal dynein arms, and is found only in organisms with motile cilia and flagella. Mutations in this gene are associated with primary ciliary dyskinesia-18, a disorder characterized by abnormalities of motile cilia. Alternatively spliced transcript variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dynein (axonemal) assembly factor 5; Dynein axonemal assembly factor 5; dynein, axonemal, assembly factor 5; HEAT repeat containing 2; HEAT repeat-containing protein 2
Gene Aliases: CILD18; DNAAF5; HEATR2
UniProt ID: (Human) Q86Y56
Entrez Gene ID: (Human) 54919, (Rat) 304332
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support