Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MEDRRAEKSC EQACESLKRQ DYEMALKHCT EALLSLGQYS MADFTGPCPL EIERIKIESL LYRIASFLQL KNYVQADEDC RHVLGEGLAK GEDAFRAVLC
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Down-regulated in human cancers protein; helicase with zinc finger domain; Probable helicase with zinc finger domain
Gene Aliases: DHRC; DRHC; HELZ; HUMORF5; KIAA0054
UniProt ID: (Human) P42694
Entrez Gene ID: (Human) 9931
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support