Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
N5-glutamine methyltransferase responsible for the methylation of the glutamine residue in the universally conserved GGQ motif of the mitochondrial translation release factor MTRF1L. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HEMK homolog 7kb; HemK methyltransferase family member 1; M.HsaHemKP; MTRF1L release factor glutamine methyltransferase; testis secretory sperm-binding protein Li 225n
Gene Aliases: 2310008M14Rik; AW049265; HEMK; HEMK1; MTQ1; RGD1308293
UniProt ID: (Human) Q9Y5R4, (Mouse) Q921L7
Entrez Gene ID: (Human) 51409, (Rat) 300989, (Mouse) 69536
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support