Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MELWGRMLWA LLSGPGRRGS TRGWAFSSWQ PQPPLAGLSS AIELVSHWTG VFEKRGIPEA RESSEYIVAH VLGAKTFQSL RPALWTQPLT SQQLQCIREL SSRRLQRMPV QYILGEWDFQ GLSLRMVPPV FIPRPETEEL VEWVLEEVAQ RSHAVGSPGS PLILEVGCGS GAISLSLLSQ LPQSRVIAVD KREAAISLTH ENAQRLRLQD RIWIIHLDMT SERSWTHLPW GPMDLIVSNP PYVFHQDMEQ LAPEIRSYED PAALDGGEEG MDIITHILAL APRLLKDSGS IFLEVDPRHP ELVSSWLQSR PDLYLNLVAV RRDFCGRPRF LHIRRSGP
N5-glutamine methyltransferase responsible for the methylation of the glutamine residue in the universally conserved GGQ motif of the mitochondrial translation release factor MTRF1L. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HEMK homolog 7kb; HemK methyltransferase family member 1; M.HsaHemKP; MTRF1L release factor glutamine methyltransferase; testis secretory sperm-binding protein Li 225n
Gene Aliases: HEMK; HEMK1; MTQ1
UniProt ID: (Human) Q9Y5R4
Entrez Gene ID: (Human) 51409
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support